missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Ammonium Test Strip Kits

Product Code. 10587302 Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
Bright Yellow to Orange
Unit Size:
81g
This item is not returnable. View return policy

Product Code. 10587302

Brand: Macherey-Nagel™ 91315

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allows for the quick and easy determination of ammonium in solutions. Macherey Nagel™ Quantofix™ Ammonium Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

  • Ready to use
  • Rapid: provides reliable results within 10 seconds
  • Excellent for industrial effluents and fertilizer-heavy rural farming areas
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Specifications

Color Indicator Bright Yellow to Orange
Format Strip
For Use With (Application) Determination of Ammonium
Graduations 0, 10, 25, 50, 100, 200, 400mg /L NH4+
Quantity 100 Pack
Storage Requirements 30°C
Test Time 10 sec.
Type Semi-qualitative Test Strip
compliance-icons
Product Identifier
  • QUANTOFIX Ammonium
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.