missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615

Product Code. 10320415 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
82 mm
85 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10320415

Brand: Macherey-Nagel™ 431003

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Widely used for routine separations.

  • Qualitative filter papers widely used for routine separations
  • Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
  • Most widely used general purpose filter paper with medium retention and flow rate
  • This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids

Specifications

Format Disc
Grade MN 615
Quantity 100/Pk.
Diameter (Metric) 30 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.