missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN QIAcard FTA Mini
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
£117.00 - £423.00
Specifications
Description | Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards. |
---|---|
Sample Type | Blood |
For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology |
No. of Samples | 2 |
Product Type | FTA Mini Card |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11969075
|
QIAGEN
WB120355 |
25 Pk. |
£117.00
Pack of 25 |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11999424
|
QIAGEN
WB120055 |
100 Pk. |
£423.00
Pack of 100 |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
- QIAcard FTA Mini format with 2 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
- FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards. | |
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology | |
FTA Mini Card |
Blood | |
2 | |
125 μL |