Learn More
QIAGEN QIAcard FTA Mini
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
Brand:  QIAGEN WB120055
Code : 69
Additional Details : Weight : 0.21000kg
 View more versions of this product
Product Code. 11999424
  • £423.00 / Pack of 100
Estimated Shipment: 05-07-2024
to see stock.
Description
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
  • QIAcard FTA Mini format with 2 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Specifications
Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards.
Blood
2
100 Pk.
Mini
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
FTA Mini Card
125 μL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
SDS
Documents
Documents
Certificates
View accessory products