Learn More
Izon qEVsingle Single-use Columns
Description
- Smallest qEV column, the qEVsingle is designed to separate out Extracellular Vesicles (EV’s) from protein and lipids in small volume samples of biological fluids
- Intended for single use, they are perfect for working with potentially biohazardous samples, or to avoid contamination when DNA/RNA testing is required. They can also be used for any small volume samples where minimal dilution and maximum recovery is required. Loading a 150μL sample results in a void volume of 1mL and EV fractions of 0.5mL
Specifications
Specifications
| Description | Izon qEVsingle |
| Column Type | Size Exclusion Column |
| For Use With (Application) | EV/exsosome isolation |