Learn More
Izon qEVsingle Rack
Click to view available options
:
For use with qEVsingle columns
Description
- The qEV Rack makes running qEV columns faster and more convenient
- It allows you to rinse and wash two columns while collecting EV fractions from another
- Wash and void volumes are collected in 15mL tubes at the bottom of the rack and EV fractions are collected into 1.5mL or 0.5mL micro tubes
- 15mL buffer hoppers attach to the top of the columns and aid in column rinsing and washing
Specifications
Specifications
| Description | Izon qEVsingle Rack |
| Column Type | Size Exclusion Column |
| For Use With (Application) | EV/exsosome isolation |