missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Carboxylase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16588-1-AP-20UL
This item is not returnable.
View return policy
Description
Pyruvate Carboxylase requires biotin and ATP to catalyse the carboxylation of pyruvate to oxaloacetate. The active enzyme is a homotetramer arranged in a tetrahedron which is located exclusively in the mitochondrial matrix. Pyruvate carboxylase is involved in gluconeogenesis, lipogenesis, insulin secretion and synthesis of the neurotransmitter glutamate. Mutations in this gene have been associated with pyruvate carboxylase deficiency.Specifications
| Pyruvate Carboxylase | |
| Polyclonal | |
| Unconjugated | |
| PC | |
| PC, PCB, pyruvate Carboxylase, Pyruvic carboxylase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18517, 25104, 5091 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11498, P52873, Q05920 | |
| Pc | |
| Pyruvate Carboxylase Fusion Protein Ag8857 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction