missing translation for 'onlineSavingsMsg'
Learn More

Pyruvate Carboxylase Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£161.00 - £375.00

Specifications

Antigen Pyruvate Carboxylase
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:500 - 1:1000
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18693821
View Documents
Novus Biologicals
NBP2-93781-0.02ml
0.02 mL
£161.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18698210
View Documents
Novus Biologicals
NBP2-93781-0.1ml
0.1 mL
£375.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pyruvate Carboxylase Polyclonal antibody specifically detects Pyruvate Carboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications

Specifications

Pyruvate Carboxylase
Western Blot, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Lipid and Metabolism
PBS (pH 7.3), 50% glycerol
5091
IgG
Affinity purified
Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:500 - 1:1000
Polyclonal
Purified
RUO
Human, Mouse, Rat
EC 6.4.1, EC 6.4.1.1, PCBPyruvic carboxylase, pyruvate carboxylase, pyruvate carboxylase, mitochondrial
Recombinant fusion protein containing a sequence corresponding to amino acids 879-1178 of human Pyruvate Carboxylase (NP_001035806.1). HSMGLGSKFKEVKKAYVEANQMLGDLIKVTPSSKIVGDLAQFMVQNGLSRAEAEAQAEELSFPRSVVEFLQGYIGVPHGGFPEPFRSKVLKDLPRVEGRPGASLPPLDLQALEKELVDRHGEEVTPEDVLSAAMYPDVFAHFKDFTATFGPLDSLNTRLFLQGPKIAEEFEVELERGKTLHIKALAVSDLNRAGQRQVFFELNGQLRSILVKDTQAMKEMHFHPKALKDVKGQIGAPMPGKVIDIKVVAGAKVAKGQPLCVLSAMKMETVVTSPMEGTVRKVHVTKDMTLEGDDLILEIE
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.