missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Carboxylase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93781-0.02ml
This item is not returnable.
View return policy
Description
Pyruvate Carboxylase Polyclonal antibody specifically detects Pyruvate Carboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Pyruvate Carboxylase | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:500 - 1:1000 | |
| EC 6.4.1, EC 6.4.1.1, PCBPyruvic carboxylase, pyruvate carboxylase, pyruvate carboxylase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 879-1178 of human Pyruvate Carboxylase (NP_001035806.1). HSMGLGSKFKEVKKAYVEANQMLGDLIKVTPSSKIVGDLAQFMVQNGLSRAEAEAQAEELSFPRSVVEFLQGYIGVPHGGFPEPFRSKVLKDLPRVEGRPGASLPPLDLQALEKELVDRHGEEVTPEDVLSAAMYPDVFAHFKDFTATFGPLDSLNTRLFLQGPKIAEEFEVELERGKTLHIKALAVSDLNRAGQRQVFFELNGQLRSILVKDTQAMKEMHFHPKALKDVKGQIGAPMPGKVIDIKVVAGAKVAKGQPLCVLSAMKMETVVTSPMEGTVRKVHVTKDMTLEGDDLILEIE | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 5091 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction