missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Pyronin Y

Product Code. 16478916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478916 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478916 Supplier MedChemExpress Supplier No. HYD0971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyronin Y (Pyronine G) is a cationic dye that intercalates RNA and has been used to target cell structures including RNA, DNA and organelles.

Pyronin Y forms fluorescent complexes with double-stranded nucleic acids (especially RNA) enabling semi-quantitative analysis of cellular RNA. Pyronin Y can be used to identify specific RNA subspecies of ribonuclear proteins complexes in live cells.

Specifications

Content And Storage -20°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (82.56 mM; Need ultrasonic) ∣ H2O : 4 mg/mL (13.21 mM; Need ultrasonic)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 302.8 g/mol
CAS 92-32-0
Synonym Pyronine G ∣ C.I. 45005
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.