Learn More
Pyrex™ Roux Culture Off-Set Neck Flasks
Ideal for cell culture
Brand: Pyrex 1350/04M
Code : NEW
Additional Details : Weight : 3.80000kg
Description
- Manufactured from chemically inert Pyrex glass for a contamination-free environment
- Withstands repeated autoclaving /li>
Specifications
600 mL | |
Advocated for cell culture |