missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Funnel with Sintered Glass Disc

Product Code. 12623212
Change view
Click to view available options
Capacity (Metric):
20 mL
30 mL
Porosity:
2
3
4
Diameter (Metric) Disc:
20mm
30mm
Height (Metric):
130 mm
144 mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. Capacity (Metric) Porosity Diameter (Metric) Disc Height (Metric) unitSize
12623212 20 mL 4 20mm 130 mm Pack of 5
11313674 20 mL 3 20mm 130 mm Each
12613212 20 mL 3 20mm 130 mm Pack of 5
11323674 20 mL 4 20mm 130 mm Each
11333674 30 mL 3 30mm 144 mm Each
15507720 20 mL 2 20mm 130 mm Pack of 10
6 options
This item is not returnable. View return policy

Product Code. 12623212

Brand: Pyrex 3730/08

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper.

Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper

  • Sintered disc manufactured from Pyrex™ borosilicate glass
  • For the filtration of solutions: - No requirement for filter paper - Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Available in a choice of porosities
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 20 mL
Diameter (Metric) Stem 8mm
Material Pyrex Glass, Borosilicate Glass
Height (Metric) 130 mm
For Use With (Application) For filtration of solutions
Diameter (Metric) Disc 20mm
Includes Sintered glass disc
Length (Metric) Stem 75mm
Porosity 4
Pore Size 10 to 16μm
Product Type Filter funnel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.