Learn More
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask
Manufactured from Pyrex™ glass for high resistance to thermal shock
£126.00 - £313.78
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10427230
|
Pyrex
1080/08D |
250 mL |
£160.00
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
12539095
|
Pyrex
1080/10D |
500 mL |
£126.00
Pack of 10 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
12549095
|
Pyrex
1080/12D |
1000 mL |
£262.00
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
12423431
|
Pyrex
1080/14D |
2000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15547660
|
Pyrex
1080/02M |
50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Complies with ISO 24450
- With wide neck to facilitate easy addition of solid samples etc
- Flat base for standing on benches and hotplates etc
- Manufactured from Pyrex™ glass for high resistance to thermal shock
- Excellent chemical durability for contamination free performance
Specifications
ISO 24450 |