Learn More
PYREX™ 95mm Diameter Glass Buchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 95mm Glass Buchner Filter Funnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
Brand: Pyrex 3710/10
Code : NEW
Additional Details : Weight : 0.44000kg
Description
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 95mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
Buchner filter funnel | |
19mm | |
250 mm | |
95mm | |
115mm | |
1 to 16μm |
500 mL | |
Pyrex Glass, Borosilicate Glass | |
For easy filtering solutions | |
Integrated filter plate sintered Glass disc | |
5 |