missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ 60mm Diameter Glass Büchner Filter Funnel

Product Code. 12613222
Change view
Click to view available options
Porosity:
0
1
2
3
4
5
Pore Size:
1.0 to 1.6μm
10 to 16μm
100 to 160μm
16 to 40μm
160 to 250μm
40 to 100μm
Unit Size:
Pack of 10
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Porosity Pore Size unitSize
12613222 1 100 to 160μm Pack of 5
11317994 2 40 to 100μm Pack of 5
11900955 0 160 to 250μm Pack of 5
11981025 3 16 to 40μm Pack of 5
12643222 4 10 to 16μm Pack of 5
15567710 5 1.0 to 1.6μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12613222 Supplier Pyrex Supplier No. 3700/02
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 60mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.

  • Comprehensive range of Büchner filter funnels with integral sintered glass discs
  • Disc and funnel body manufactured from chemically resistant Pyrex™ glass
  • For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
  • Choice of disc diameters and porosities
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Diameter (Metric) Stem 13mm
Material Borosilicate Glass
Height (Metric) 180 mm
For Use With (Application) For convenient filtration of solutions
Diameter (Metric) Disc 60mm
Includes Integrated filter plate sintered Glass disc
Length (Metric) Stem 100mm
Porosity 1
Pore Size 100 to 160μm
Product Type Buchner Funnel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.