missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ 40mm Diameter Glass Büchner Filter Funnel

Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 40mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.

Brand:  Pyrex 3630/10

Code : A0

Additional Details : Weight : 0.48000kg

 View more versions of this product

Product Code. 12602142

  • £203.30 / Pack of 5
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

  • Comprehensive range of Büchner filter funnels with integral sintered glass discs
  • Disc and funnel body manufactured from chemically resistant Pyrex™ glass
  • For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
  • Choice of disc diameters and porosities
Specifications

Specifications

Buchner filter funnel
8mm
170 mm
40mm
100mm
1.0 to 1.6μm
50 mL
Pyrex Glass, Borosilicate Glass
For convenient filtration of solutions
Integrated filter plate sintered Glass disc
5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers