missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXMP4 Antibody (7S41) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£153.00 - £426.00
Specifications
| Antigen | PXMP4 |
|---|---|
| Clone | 7S41 |
| Concentration | LYOPH |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18107854
|
Novus Biologicals
NBP2-21987-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159251
|
Novus Biologicals
NBP2-21987 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PXMP4 Monoclonal specifically detects PXMP4 in Mouse samples. It is validated for ELISA.Specifications
| PXMP4 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| P21245 | |
| 11264 | |
| IgG2 | |
| Protein G purified |
| 7S41 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa | |
| PXMP4 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title