missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXMP4 Antibody (7S41) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-21987
This item is not returnable.
View return policy
Description
PXMP4 Monoclonal specifically detects PXMP4 in Mouse samples. It is validated for ELISA.
Specifications
| PXMP4 | |
| Monoclonal | |
| LYOPH | |
| ELISA 1:1000-1:2000 | |
| P21245 | |
| PXMP4 | |
| Mouse recombinant PXMP4 | |
| 0.1 mg | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
| ELISA | |
| 7S41 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa | |
| Rat | |
| Protein G purified | |
| RUO | |
| 11264 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction