missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PWP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PWP2 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16848433
|
Proteintech
11390-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838433
|
Proteintech
11390-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MW of this protein is 102 kDa, and this antibody specifically recognizes the 102 kDa protein.
PWP2 belongs to the WD repeat PWP2 family. It contains 14 WD repeats. The exact function of PWP2 is not known.Specifications
| PWP2 | |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q15269 | |
| 5822 | |
| PWP2 Fusion Protein Ag1936 | |
| Primary | |
| -20°C | |
| PWP2 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| EHOC 17, PWP2, PWP2H | |
| PWP2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title