missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PWP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11390-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 102 kDa, and this antibody specifically recognizes the 102 kDa protein.
PWP2 belongs to the WD repeat PWP2 family. It contains 14 WD repeats. The exact function of PWP2 is not known.Specifications
| PWP2 | |
| Polyclonal | |
| Unconjugated | |
| PWP2 | |
| EHOC 17, PWP2, PWP2H | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5822 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15269 | |
| PWP2 | |
| PWP2 Fusion Protein Ag1936 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction