missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ PuroSorb™ PAD Macroporous Adsorbents
Click to view available options
Quantity:
250 g
1 kg
5 kg
Product Line:
PAD1200
PAD610
PAD950
Unit Size:
1kg
250g
5kg
Description
- Stable in acidic or alkaline solutions and in most organic solvents
- Regenerate under mild conditions for prolonged life
- Chemically stable, insoluble
- Spherical beads
- Macroporous structure
- Made of styrene, DVB or methacrylate
- Stevia Purification
- Peptide extraction
- Decolorization
- Macrolide purification
- Compliant with ResAP(2004) 3
- Halal and TSE/BSE/GMO free
- Compliant with FDA regulation 21 CFR 173.65
Specifications
Specifications
| Final Product Type | White to off-white spherical beads |
| Product Line | PAD610 |
| Product Type | Macroporous Polymethacrylic/DVB Adsorbent |
| Quantity | 250 g |
| pH Range | 1 to 14 pH |
| Particle Size | 350 to 1200 μm |
| Content And Storage | Use within 5 years from date of manufacture, store at 2°C to 20°C |
| Format | Wet |
| Moisture | 60 to 71% |
| Porosity | 50 Å |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction