missing translation for 'onlineSavingsMsg'
Learn More

Puromycin Dihydrochloride, TRC

Product Code. 30428462
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30428462 25 mg 25mg
30411561 250 mg -
2 options
This item is not returnable. View return policy

Product Code. 30428462

Brand: Toronto Research Chemicals TRCP84032025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-58-2
Formula Weight 543.1764
InChI Formula InChI=1S/C22H29N7O5.2ClH/c1-28(2)19-17-20(25-10-24-19)29(11-26-17)22-18(31)16(15(9-30)34-22)27-21(32)14(23)8-12-4-6-13(33-3)7-5-12;;/h4-7,10-11,14-16,18,22,30-31H,8-9,23H2,1-3H3,(H,27,32);2*1H/t14-,15+,16+,18+,22+;;/m0../s1
IUPAC Name (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
Molecular Formula C22 H29 N7 O5 . 2 Cl H
Molecular Weight (g/mol) 544.43
SMILES Cl.Cl.COc1ccc(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)n3cnc4c(ncnc34)N(C)C)cc1
Synonym 3'-[[(2S)-2-amino-3-(4-methoxyphenyl)-1-oxopropyl]amino]-3'-deoxy-N,N-dimethyl-adenosine hydrochloride (1:2), Puromycin dihydrochloride, Stylomycin dihydrochloride
Chemical Name or Material Puromycin hydrochloride

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.