missing translation for 'onlineSavingsMsg'
Learn More

DURAN™ PURE Silicone PTFE-coated Seal
BIOPHARMA

Product Code. p-7182007 Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 25
GL 45
GLS 80
Unit Size:
Pack of 10

Product Code. 16424954

Brand: DURAN™ 292481456

Only null left
Add to basket
Edge
Add to basket

High-quality silicone seals developed for the needs of the pharmaceutical and biotech industry

  • High-quality silicone seals developed for the needs of the pharmaceutical and biotech industry
  • High-quality, PTFE coated silicone seal for use with DURAN™ PURE Premium Screw Cap, GL 25
  • Packaged in clean room
  • Single batch delivery
  • Certificate of Analysis (CoA)
  • Visual and dimensional inspection
  • Change control management and information of customer in case of changes
  • USP <88> Class VI Pharmaceutical grade materials
TRUSTED_SUSTAINABILITY

Specifications

Color Light Ivory, Gray
Description PTFE-coated silicone seal
Material Platinum-cured Silicone, PTFE
Thickness (Metric) 3 mm
Thread Size (Metric) GL 25
For Use With (Equipment) GL 25 cap
Diameter (Metric) 23.5 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.