missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PUMA alpha Recombinant Rabbit Monoclonal Antibody (23GB1935), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen PUMA alpha
Clone 23GB1935
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088016
GREENER_CHOICE
View Documents
Invitrogen™
MA552466
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089676
View Documents
Invitrogen™
MA552467
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PUMA alpha Recombinant Monoclonal antibody specifically detects PUMA alpha in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

PUMA alpha
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
Q80ZG6, Q99ML1, Q9BXH1
170770, 27113, 317673
IgG
Affinity chromatography
Antibody
23GB1935
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; FLJ42994; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1
A synthesized peptide derived from human PUMA (150-193AA).
Primary
-20°C
BBC3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.