Learn More
Invitrogen™ PUMA alpha Recombinant Rabbit Monoclonal Antibody (23GB1935), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552467
 View more versions of this product
Product Code. 30089676
  • £162.00 / 20µL
Description
PUMA is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMA-a and PUMA-b. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis. Antisense inhibition of PUMA expression reduced the apoptotic response to p53, and PUMA is likely to play a role in mediating p53-induced cell death through the cytochrome c/Apaf-1-dependent pathway.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PUMA alpha
Recombinant Monoclonal
Unconjugated
BBC3
BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; FLJ42994; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1
Rabbit
Affinity chromatography
RUO
170770, 27113, 317673
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1935
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q80ZG6, Q99ML1, Q9BXH1
BBC3
A synthesized peptide derived from human PUMA (150-193AA).
20 μL
Primary
Human, Mouse, Rat