Learn More
PUF60 Mouse, anti-Human, Clone: 1A3A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68533-1-IG-20UL
Description
The PUF60 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PUF60. This antibody recognizes human, mouse, rat, rabbit antigen. The PUF60 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PUF60 | |
| 1A3A1 | |
| FBP interacting repressor, FIR, FLJ31379, PUF60, Ro binding protein 1, RoBP1, RoBPI, Siah binding protein 1, Siah BP1, SIAHBP1 | |
| 20 μL | |
| 22827 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 150 kDa | |
| IgG1 |