missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUF60 Mouse, anti-Human, Clone: 1A3A1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PUF60 |
|---|---|
| Clone | 1A3A1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006195
|
Proteintech
68533-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18036905
|
Proteintech
68533-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PUF60 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PUF60. This antibody recognizes human, mouse, rat, rabbit antigen. The PUF60 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PUF60 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Rabbit | |
| FBP interacting repressor, FIR, FLJ31379, PUF60, Ro binding protein 1, RoBP1, RoBPI, Siah binding protein 1, Siah BP1, SIAHBP1 | |
| Primary |
| 1A3A1 | |
| Western Blot, ELISA | |
| Mouse | |
| 22827 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title