missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | PTPN6 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058305
|
Proteintech
CL488-24546-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18099355
|
Proteintech
CL488-24546-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PTPN6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PTPN6. This antibody recognizes human antigen. The PTPN6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PTPN6 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| HCP, HCPH, HPTP1C, PTP 1C, PTP1C, PTPN6, SH PTP1, SHP 1, SHP 1L, SHP1 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 5777 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title