Learn More
PTPN6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-24546-100UL
Description
The PTPN6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PTPN6. This antibody recognizes human antigen. The PTPN6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PTPN6 | |
| 1000 μg/mL | |
| HCP, HCPH, HPTP1C, PTP 1C, PTP1C, PTPN6, SH PTP1, SHP 1, SHP 1L, SHP1 | |
| 100 μL | |
| 5777 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.