missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £366.00
Specifications
| Antigen | PTPN4 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666282
|
Novus Biologicals
NBP2-94872-0.02ml |
0.02 mL |
£159.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695822
|
Novus Biologicals
NBP2-94872-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTPN4 Polyclonal antibody specifically detects PTPN4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PTPN4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.3), 50% glycerol | |
| 5775 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.1.3.48, MEG, megakaryocyte phosphatase, megakaryocyte protein-tyrosine phosphatase, protein tyrosine phosphatase MEG1, protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte), Protein-tyrosine phosphatase MEG1, PTPase-MEG1, PTPMEG, PTPMEG1, tyrosine-protein phosphatase non-receptor type 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human PTPN4 (NP_002821.1). MTSRFRLPAGRTYNVRASELARDRQHTEVVCNILLLDNTVQAFKVNKHDQGQVLLDVVFKHLDLTEQDYFGLQLADDSTDNPRWLDPNKPIRKQLKRGSPYSLNFRVKFFVSDPNKLQEEYTRYQYFLQIKQDILTGRLPCPSNTAALLASFAVQSELGDYDQSENLSGYLSDYSFIPNQPQDFEKEIAKLHQQHIGLSPAEAEFNYLNTARTLELYGVEFHYARDQSNNEIMIGVMSGG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title