missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94872-0.1ml
This item is not returnable.
View return policy
Description
PTPN4 Polyclonal antibody specifically detects PTPN4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PTPN4 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| EC 3.1.3.48, MEG, megakaryocyte phosphatase, megakaryocyte protein-tyrosine phosphatase, protein tyrosine phosphatase MEG1, protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte), Protein-tyrosine phosphatase MEG1, PTPase-MEG1, PTPMEG, PTPMEG1, tyrosine-protein phosphatase non-receptor type 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human PTPN4 (NP_002821.1). MTSRFRLPAGRTYNVRASELARDRQHTEVVCNILLLDNTVQAFKVNKHDQGQVLLDVVFKHLDLTEQDYFGLQLADDSTDNPRWLDPNKPIRKQLKRGSPYSLNFRVKFFVSDPNKLQEEYTRYQYFLQIKQDILTGRLPCPSNTAALLASFAVQSELGDYDQSENLSGYLSDYSFIPNQPQDFEKEIAKLHQQHIGLSPAEAEFNYLNTARTLELYGVEFHYARDQSNNEIMIGVMSGG | |
| 0.1 mL | |
| Protein Phosphatase | |
| 5775 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Übermitteln Sie eine inhaltliche Korrektur