missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN14/PTPD2 Antibody (2B3H7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | PTPN14/PTPD2 |
|---|---|
| Clone | 2B3H7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256904
|
Novus Biologicals
NBP2-61809 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600487
|
Novus Biologicals
NBP2-61809-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTPN14/PTPD2 Monoclonal specifically detects PTPN14/PTPD2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PTPN14/PTPD2 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| EC 3.1.3, EC 3.1.3.48, MGC126803, PEZcytoskeletal-associated protein tyrosine phosphatase, protein tyrosine phosphatase, non-receptor type 14, Protein-tyrosine phosphatase pez, PTP36, tyrosine-protein phosphatase non-receptor type 14 | |
| PTPN14 | |
| IgG1 | |
| Protein G purified |
| 2B3H7 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| 5784 | |
| Purified recombinant fragment of human PTPN14 (AA: 896-1169) expressed in E. Coli. | |
| Primary | |
| 135.3 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title