missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN14/PTPD2 Antibody (2B3H7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61809
This item is not returnable.
View return policy
Description
PTPN14/PTPD2 Monoclonal specifically detects PTPN14/PTPD2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PTPN14/PTPD2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| EC 3.1.3, EC 3.1.3.48, MGC126803, PEZcytoskeletal-associated protein tyrosine phosphatase, protein tyrosine phosphatase, non-receptor type 14, Protein-tyrosine phosphatase pez, PTP36, tyrosine-protein phosphatase non-receptor type 14 | |
| Mouse | |
| 135.3 kDa | |
| 0.1 mL | |
| 5784 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2B3H7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PTPN14 | |
| Purified recombinant fragment of human PTPN14 (AA: 896-1169) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction