Learn More
GeneBLAzer™ PTGER1-NFAT-bla CHO-K1 Cells
GeneBLAzer™ PTGER1-NFAT-bla CHO-K1 Cells contain the human Prostaglandin E Receptor 1 (Subtype 1).
Brand: GeneBLAzer™ K1851
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: PTGER1
- Accession Number: NM_000955.2
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ PTGER1-NFAT-bla CHO-K1 cells contain the human Prostaglandin E Receptor 1 (Subtype 1) (PTGER1), (Accession # NM_000955.2) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. no. K1534) contain a beta-lactamase reporter gene under control of the NFAT Response Element. GeneBLAzer™ PTGER1-NFAT-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of PGE2 (Figure 1). In addition, GeneBLAzer™ PTGER1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Liquid Nitrogen | |
GPCRs | |
8 x 106 Cells |
Fluorescent | |
PTGER1-NFAT-bla CHO-K1 cells, PTGER1-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
Dividing Cells |