Learn More
Cowie Technology™ PTFE T-Type Connector
Accepts any rigid or semi-rigid tubing. Cowie Technology™ PTFE T-Type Connector locks in place quickly with a bayonet type coupling.
Brand:  Cowie Technology™ 016.1104.9.2
Additional Details : Weight : 0.01400kg
 View more versions of this product
Product Code. 10591622
  • £12.70 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Bayonet fitting
  • For use with rigid and semi rigid tubing such as glass, FEP or PTFE
Specifications
Specifications
PTFE T-Type Connector
Autoclavable
PTFE
-200°C
33.5 mm
White
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
9 mm
51.5 mm
1 Each
16 mm
280°C