Learn More
Cowie Technology™ PTFE T-Type Connector
Accepts any rigid or semi-rigid tubing. Cowie Technology™ PTFE T-Type Connector locks in place quickly with a bayonet type coupling.
Brand:  Cowie Technology™ 016.1103.7.2
Additional Details : Weight : 0.00900kg
 View more versions of this product
Product Code. 10437552
  • £11.60 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Bayonet fitting
  • For use with rigid and semi rigid tubing such as glass, FEP or PTFE
Specifications
Specifications
PTFE T-Type Connector
Autoclavable
PTFE
Flexible tubing in PTFE, Viton, silicone rubber or similar materials
1 Each
14 mm
280°C
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
6.8 mm
44 mm
-200°C
29 mm
White