missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE Square-End Tweezers

Product Code. 10281802 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
100 mm
150 mm
200 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10281802 200 mm Each
10510292 150 mm Each
10282092 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10281802
Only null left
Add to basket
Add to basket

Cowie Technology™ PTFE Square-End Tweezers are inert to virtually all chemical reagents and completely non-contaminating.

  • Chemically inert
  • Non-contaminating
  • Usable to 280°C
  • Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals

Specifications

Tip Style Square
No. per Pack 1 Each
Material PTFE
Length (Metric) 200 mm
Product Type PTFE Forceps / Tweezers
Finish Smooth
Height (Metric) Body 12 mm
Thickness 12 mm
Width (Metric) Body 25 mm
Width (Metric) Tip 11 mm
Thickness (Metric) Tip 5 mm
Dimensions (L x W) 200 x 25 mm
Width (Metric) 25 mm
Style Rigid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.