missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Insert for Screw Cap GL 14, Silicone/PTFE (Screw Connection Cap GL 45 Accessory)

Product Code. 10399652
Click to view available options
Diameter (Metric) Inner:
ID 1.6 mm
ID 3.0 mm
ID 3.2 mm
ID 6.0 mm

Accessory for 2 or 3 port GL 45 connection cap system

Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.

  • Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts
  • Available with 2, or 3 threaded outlets (ports)
  • Five different tubing diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be used by changing the inserts
  • By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap
  • Fully compatible with all the GL 45 threads and therefore highly versatile
  • Fully compatible with all the GLS 80 threads and therefore highly versatile

Suitable Accessory: 1129814

TRUSTED_SUSTAINABILITY

Specifications

Description Insert for screw cap GL 14, ID 4.0 mm (∼1/4 inch)
Diameter (Metric) Inner ID 6.0 mm
For Use With (Application) Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply
Material Polypropylene, PTFE
Quantity 1 Ea.
Thread Size (Metric) GL 14
Autoclavable Autoclavable
Diameter (Metric) 87 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'