Learn More
Cole-Parmer™ PTFE Screen Gaskets
Protect your pump, valves and other components in fluid line
Brand:  Cole-Parmer™ 3053804
Additional Details : Weight : 0.02270kg
 View more versions of this product
Product Code. 16679275
  • £167.00 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Specifications
Specifications
10
PTFE Gasket
PTFE
2.54 cm