Learn More
Cole-Parmer™ PTFE Screen Gaskets
Description
- Pre-filter before critical membrane filters
- Generate turbulence for enhanced mass or heat transfer
- Purify in chromatography applications
Specifications
Specifications
| Mesh Size | 100 |
| Diameter (Metric) Outside | 5.08 cm |
| Material | PTFE |
| Type | Gasket |