Learn More
Cole-Parmer™ PTFE Screen Gaskets
Product Code. 16629295
Change view
Click to view available options
Diameter (Metric) Outside:
1.27 cm
2.54 cm
3.81 cm
5.08 cm
Mesh Size:
4
10
20
40
100
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Mesh Size unitSize
16629295 1.27 cm 100 Each
16679275 2.54 cm 10 Each
16659285 2.54 cm 4 Each
16689275 3.81 cm 10 Each
16639295 3.81 cm 100 Each
16699275 5.08 cm 10 Each
16639285 5.08 cm 20 Each
16679285 5.08 cm 40 Each
16649295 5.08 cm 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 16629295 Supplier Cole-Parmer™ Supplier No. 3053880
Protect your pump, valves and other components in fluid line
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Specifications
Mesh Size 100
Diameter (Metric) Outside 1.27 cm
Material PTFE
Type Gasket