missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Right Angle Connector

Product Code. 16295971 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
8 mm
Unit Size:
Pack of 5

Product Code. 16295971

Only null left
Add to basket
Add to basket

Fisherbrand™ PTFE Right Angle Connectors with bayonet fittings are for use with flexible tubing in PTFE, VITON, Silicone rubber or similar materials.

  • Bayonet Fitting
  • Pure PTFE
  • Can be sterlised at 135°C
  • Total Chemical Resistance
  • High Purity
  • Non contaminating
  • Easy to clean

Specifications

Description Fisherbrand™ PTFE Right Angle Connector, Arm Diameter 8mm x Bore 3mm
Diameter (Metric) 8 mm
Material PTFE
For Use With PTFE, VITON, Silicone rubber Tubing
Port Size 3 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.