missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH PTFE Grinding Jars
£901.00 - £960.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10296792
|
RETSCH
01.462.0244 |
35 mL |
£901.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
14831631
|
RETSCH
01.462.0335 |
25 mL |
£960.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Specifications
Grinding Jar | |
PTFE |