missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Screw-top Bottle

Product Code. 10762741 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
1 mL
Unit Size:
Each

Product Code. 10762741

Only null left
Add to basket
Add to basket

Molded-in hand grips provide ease of manipulation. The PTFE screw-top bottle is chemically resistant and inert.

  • Isostatically molded in pure PTFE and with super smooth internal finish, FDA approved
  • Insoluble and non-contaminating
  • Exceptional thermal resistance can be used from -200°C to +280°C
  • Hand grips for ease of manipulation
  • Do not use, when sealed, for chemical reactions or digestions, loosen lid when thermal sterilizing

Specifications

Capacity (Metric) 1 mL
Color White
Diameter (Metric) Inner Neck 10mm
For Use With (Application) Storage of hazardous chemicals
Height (Metric) With Closure 27mm
Material PTFE
Max. Temperature (Metric) 280°C
Temperature Range (Metric) -200 to +280°C
Thickness (Metric) 2mm
Weight (Metric) 8g
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Closure Included Yes
Graduated No
Product Type Storage Bottle
Diameter (Metric) 16 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.