Learn More
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrers are for use with plain glass, metal or PTFE Shafts.
Brand: Fisherbrand™
Additional Details : Weight : 0.15000kg
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer | |
16 mm | |
-200°C to +280°C |
100 mm | |
1 | |
PTFE |