Learn More
Masterflex™ PTFE 2-Way Direct Lift Solenoid Valve
Product Code. 11759759
Change view
Click to view available options
Material:
Teflon
Unit Size:
Each
Product Code. Material unitSize
11759759 Teflon Each
1 options
Product Code. 11759759 Supplier Masterflex™ Supplier No. 0154009
Estimated Shipment: 02-03-2027
Add to basket
Add to basket
Specifications
For Use With (Application) Ideal for use with high-purity or corrosive fluids
Max. Operating Temperature (Metric) 70°C
Material Teflon
Quantity 1 Each
Type 2-way Direct Lift Solenoid Valve
Voltage 12 VDC