Learn More
PTEN Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22034-1-AP-20UL
This item is not returnable.
View return policy
Description
The PTEN antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PTEN. This antibody recognizes human, mouse antigen. The PTEN antibody has been validated for the following applications: FC, IHC, IP, WB, ELISA analysis.
Specifications
| PTEN | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 5728 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| 307 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |