missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTEN Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£133.61 - £349.17
Specifications
| Antigen | PTEN |
|---|---|
| Concentration | 307 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
The PTEN antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PTEN. This antibody recognizes human, mouse antigen. The PTEN antibody has been validated for the following applications: FC, IHC, IP, WB, ELISA analysis.
Specifications
| PTEN | |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 307 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 5728 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title