missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSTAIR Antibody (PSTAIR), Novus Biologicals™
Mouse Monoclonal Antibody
£170.00
Specifications
| Antigen | PSTAIR |
|---|---|
| Clone | PSTAIR |
| Concentration | 7.1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot |
Description
PSTAIR Monoclonal specifically detects PSTAIR in Human, Mouse, Avian, Plant, Primate, Xenopus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| PSTAIR | |
| 7.1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| EGVPSTAIREISLLKE, EGVPSTAIREISLLKE sequence, PSTAIR sequence | |
| IgG1 | |
| Unpurified | |
| PSTAIR (PSTAIR) |
| PSTAIR | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| Synthetic peptide: EGVPSTAIREISLLKE conjugated to BSA. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title