missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSTAIR Antibody (PSTAIR), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10345-0.025ml
This item is not returnable.
View return policy
Description
PSTAIR Monoclonal specifically detects PSTAIR in Human, Mouse, Avian, Plant, Primate, Xenopus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| PSTAIR | |
| Monoclonal | |
| 7.1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot | |
| PSTAIR | |
| Unconjugated | |
| EGVPSTAIREISLLKE, EGVPSTAIREISLLKE sequence, PSTAIR sequence | |
| Synthetic peptide: EGVPSTAIREISLLKE conjugated to BSA. | |
| 0.025 mL | |
| Cell Cycle and Replication | |
| PSTAIR (PSTAIR) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction